CAS RN 65405-58-5
CAS RN 65405-58-5
IUPAC Name:
1,2-diethenylbenzene; [(2-ethenylphenyl)methyl]dimethylamine
Synonyms:
[(2-ethenylphenyl)methyl]dimethylamine; divinylbenzene
Molecular Weight (g/mol):
291.44
Molecular Formula:
C21H25N
InChi Key:
OTCJDTFYONDHFM-UHFFFAOYSA-N
SMILES:
C=CC1=CC=CC=C1C=C.CN(C)CC1=CC=CC=C1C=C
1
–
1
of
1
results