CAS RN 656-65-5
CAS RN 656-65-5
IUPAC Name:
4-bromo-3-fluoroaniline
Synonyms:
4-bromo-3-fluoroaniline
Molecular Weight (g/mol):
190.02
Molecular Formula:
C6H5BrFN
InChi Key:
YTMVYYAKOPIJCZ-UHFFFAOYSA-N
SMILES:
NC1=CC=C(Br)C(F)=C1
1
–
2
of
2
results