accessibility menu, dialog, popup

UserName

CAS RN 74-82-8

CH4

CAS RN 74-82-8

IUPAC Name: methane
Synonyms: methane collocarb carbene, 13c-labeled 13C-labeled carbene kosmos spheron magecol grosafe humenegro furnal
Molecular Weight (g/mol): 16.04
Molecular Formula: CH4
InChi Key: VNWKTOKETHGBQD-UHFFFAOYSA-N
SMILES: C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results