CAS RN 760961-03-3
CAS RN 760961-03-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hederacoside D, MedChemExpress
MedChemExpress Hederacoside D is one of the bioactive saponins from Hedera helix, and plays pivotal roles in the overall biological activity.