accessibility menu, dialog, popup

UserName

CAS RN 7727-73-3

H2OH2OH2OH2OH2OH2OH2OH2OH2OH2ONaNaOSOOO

CAS RN 7727-73-3

IUPAC Name: disodium decahydrate sulfate
Synonyms: glauber's salt sodium sulphate decahydrate glaubersalz sodium sulfate decahydrate
Molecular Weight (g/mol): 322.19
Molecular Formula: H20Na2O14S
InChi Key: RSIJVJUOQBWMIM-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (12)
  • (8)
Quantity 
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
Grade 
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (7)
  • (3)
  • (2)
  • (12)
  • (8)
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (3)