CAS RN 778270-11-4
CAS RN 778270-11-4
IUPAC Name:
3-(6-{[4-(trifluoromethoxy)phenyl]amino}pyrimidin-4-yl)benzamide
Synonyms:
3-(6-{[4-(trifluoromethoxy)phenyl]amino}pyrimidin-4-yl)benzamide
Molecular Weight (g/mol):
374.32
Molecular Formula:
C18H13F3N4O2
InChi Key:
WEVYNIUIFUYDGI-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=CC(=C1)C1=CC(NC2=CC=C(OC(F)(F)F)C=C2)=NC=N1
1
–
1
of
1
results
1
GNF-2, MedChemExpress
MedChemExpress GNF-2 is a highly selective, allosteric, non-ATP competitive inhibitor of Bcr-Abl. GNF-2 inhibits Ba/F3.p210 proliferation with an IC50 of 138 nM .