CAS RN 79886-50-3
CAS RN 79886-50-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2,3,6-Tetragalloylglucose, MedChemExpress
MedChemExpress 1,2,3,6-Tetragalloylglucose is a potent UDP glucuronosyltransferase 1 family, polypeptide A1 (UGT1A1) inhibitor, with a Ki of 1.68 μM.