CAS RN 802906-73-6
CAS RN 802906-73-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Basimglurant, MedChemExpress
MedChemExpress Basimglurant (RG7090) is a potent, selective and orally available mGlu5 negative allosteric modulator with a Kd of 1.1 nM.