CAS RN 83805-11-2
CAS RN 83805-11-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Falecalcitriol, MedChemExpress
MedChemExpress Falecalcitriol(Fulstan; Hornel) is an analog of calcitriol; has a higher potency both in vivo and in vitro systems, and longer duration of action in vivo.