CAS RN 84680-54-6
CAS RN 84680-54-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Enalaprilat dihydrate, MedChemExpress
MedChemExpress Enalaprilat dihydrate (MK-422), the active metabolite of the oral prodrug Enalapril, is a potent, competitive and long-acting angiotensin-converting enzyme (ACE) inhibitor, with an IC50 of 1.94 nM. Enalaprilat dihydrate can be used for the research of hypertension.