CAS RN 878385-84-3
CAS RN 878385-84-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AZD-5069, MedChemExpress
MedChemExpress AZD-5069 is a potent CXCR2 chemokine receptor antagonist, used for caner treatment.