accessibility menu, dialog, popup

UserName

CAS RN 88122-99-0

CH3CH3OONHNNHOOCH3CH3NHNOOCH3CH3N

CAS RN 88122-99-0

IUPAC Name: 2-ethylhexyl 4-({4,6-bis[(4-{[(2-ethylhexyl)oxy]carbonyl}phenyl)amino]-1,3,5-triazin-2-yl}amino)benzoate
Synonyms: ethylhexyl triazone
Molecular Weight (g/mol): 823.09
Molecular Formula: C48H66N6O6
InChi Key: JGUMTYWKIBJSTN-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC=C(NC2=NC(NC3=CC=C(C=C3)C(=O)OCC(CC)CCCC)=NC(NC3=CC=C(C=C3)C(=O)OCC(CC)CCCC)=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results