CAS RN 925676-48-8
CAS RN 925676-48-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
LAS101057, MedChemExpress
MedChemExpress LAS101057 is a potent, selective, and orally efficacious A2B receptor antagonist.