CAS RN 94-16-6
CAS RN 94-16-6
IUPAC Name:
sodium 2-[(4-aminophenyl)formamido]acetate
Synonyms:
paraaminohippurate monosodium p-aminohippurate p-aminohippurate sodium (TN) aminohippurate sodium (TN) natrium 4-aminohippurat sodium p-aminophippurate p-aminohippurate sodium sodium 4-aminohippurate
Molecular Weight (g/mol):
216.17
Molecular Formula:
C9H9N2NaO3
InChi Key:
UNZMYCAEMNVPHX-UHFFFAOYSA-M
SMILES:
[Na+].NC1=CC=C(C=C1)C(=O)NCC([O-])=O
1
–
1
of
1
results
1
Aminohippurate sodium, MedChemExpress
MedChemExpress Aminohippurate sodium is a diagnostic agent useful in medical tests involving the kidney used in the measurement of renal plasma flow.