accessibility menu, dialog, popup

UserName

CAS RN 942425-68-5

NHOHNNHCl

CAS RN 942425-68-5

IUPAC Name: 2-(pyridin-4-yl)-1H,4H,5H,6H,7H-pyrrolo[3,2-c]pyridin-4-one hydrochloride
Synonyms: 2-(pyridin-4-yl)-1H,5H,6H,7H-pyrrolo[3,2-c]pyridin-4-one hydrochloride
Molecular Weight (g/mol): 249.7
Molecular Formula: C12H12ClN3O
InChi Key: IMVNFURYBZMFDZ-UHFFFAOYSA-N
SMILES: Cl.O=C1NCCC2=C1C=C(N2)C1=CC=NC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results