CAS RN 97540-22-2
CAS RN 97540-22-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-Adenosyl-L-methionine disulfate tosylate, MedChemExpress
MedChemExpress S-Adenosyl-L-methionine disulfate tosylate (Ademetionine disulfate tosylate) is the principal biological methyl donor synthesized in all mammalian cells but most abundantly in the liver.