Exam Gloves Frequently Asked Questions
Are Nitrile Gloves Hypoallergenic?
As nitrile does not contain any latex protein, it has a lower allergy rate than latex. Nitrile is the superior choice when it comes to people with allergies or sensitive skin.
Nitrile or Latex for Mechanics Gloves?
Nitrile gloves are stronger than latex gloves. They are also more resistant to chemicals and have higher puncture resistance.
How Are Nitrile Gloves Made?
Nitrile is a synthetic copolymer made by combining monomers such as butadiene and acrylonitrile in a big vat.
Are Nitrile Gloves Recyclable?
Nitrile gloves can be recycled. However, they need to go in a separate special recycling bin, as opposed to your standard recycling bin. As part of our sustainability program the Fisher Scientific channel is happy to introduce you to some recycling possibilities. For example, TerraCycle™ is a company that provides specialist disposable glove recycling boxes. You can buy them online, fill the box with your used disposable nitrile, vinyl or latex gloves, and simply ship the box to the collection address. Shipping is already included in the price of the box!
What are Powder-Free Nitrile Gloves?
You can find disposable gloves in both powdered and powder-free versions. Powder-free nitrile gloves are made with a powder-free formulation that reduces the risk of contact dermatitis. In addition, this type of glove can help reduce the risk of particulate contamination during use. With powdered nitrile gloves, there is a risk of cross-contamination from the powder when the gloves are removed.
What Are Vinyl Gloves?
Vinyl gloves were developed after many people started developing latex allergies due to excessive use of latex gloves. They are usually odor-free and one of their major advantages is that they can withstand all types of alcohol-based chemicals.