Request a Quote - ÄKTA
ÄKTA™ start protein purification system
ÄKTA start offers the following features:
- Compact solution for quick and reliable one-step protein purification
- Sample injection and fraction collection options
- Quick start methods and templates for common purification techniques
- Intuitive touchscreen display and real-time monitoring
Specifications:
- Flow rate: 0.5 to 5mL/min
- Pump type: peristaltic pump - single channel, four-roller pump head with low pulsation
- Max. operating pressure: 0.5MPa
- UV, fixed wavelength: Yes, 280nm
- Conductivity range: 0 to 300mS/cm
- Software: UNICORN start 1.0 or higher