missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-07047MFV610
This item is not returnable.
View return policy
Description
PDK4 Polyclonal antibody specifically detects PDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PDK4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial | |
| Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot# Q16654] | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction | |
| 5166 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction