missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A8/ZIP8 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76505CL1
This item is not returnable.
View return policy
Description
SLC39A8/ZIP8 Polyclonal antibody specifically detects SLC39A8/ZIP8 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| SLC39A8/ZIP8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| BCG induced integral membrane protein BIGM103, BIGM103BCG-induced integral membrane protein in monocyte clone 103 protein, LIV-1 subfamily of ZIP zinc transporter 6, LZT-Hs6, solute carrier family 39 (metal ion transporter), member 8, solute carrier family 39 (zinc transporter), member 8, Solute carrier family 39 member 8, zinc transporter ZIP8, ZIP8, ZIP-8, Zrt- and Irt-like protein 8 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human ZIP8. The immunogen is located within the last 50 amino acids of ZIP8. Amino Acid Squence: PEMDMLREKVTGRKTDF | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Signal Transduction | |
| 64116 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction