missing translation for 'onlineSavingsMsg'
Learn More

1,1':3',1″-Terphenyl-5'-boronic acid, 95%

Product Code. 11476600
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11476600

Brand: Thermo Scientific Alfa Aesar H27640.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128388-54-5 | C18H15BO2 | 274.126 g/mol

A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

Solubility
Insoluble in water.

Notes
Refrigerated (2-4°C). Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128388-54-5
Molecular Formula C18H15BO2
Molecular Weight (g/mol) 274.126
MDL Number MFCD09953491
InChI Key MRBZYVMZUBUDAX-UHFFFAOYSA-N
Synonym 3,5-diphenylphenyl boronic acid, 1,1':3',1-terphenyl-5'-ylboronic acid, 1,1':3',1-terphenyl-5'-boronic acid, 3,5-diphenylbenzeneboronic acid, 5'-m-terphenylboronic acid, boronic acid, 1,1':3',1-terphenyl-5'-yl, boronic acid,b-1,1':3',1-terphenyl-5'-yl, 5-phenyl-1,1'-biphenyl-3-ylboronic acid, 3,5-diphenylphenyl, acmc-1cc4x
PubChem CID 14739363
IUPAC Name (3,5-diphenylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3)(O)O

Specifications

Melting Point 292°C to 296°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 274.13
Percent Purity 95%
Chemical Name or Material 1,1':3',1''-Terphenyl-5'-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.