missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Product Code. 15444827
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15444827

Brand: Thermo Scientific Alfa Aesar H32138.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1914-65-4 | C11H12O2 | 176.215 g/mol

1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-1-naphthoic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid is a reagent used to produce protease inhibitors.

Solubility
Soluble in methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1914-65-4
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD02179127
InChI Key VDLWTJCSPSUGOA-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-1-naphthoic acid, 1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydro, tetralin-1-carboxylic acid, 1,2,3,4-tetrahedro-naphthoic acid, 1,2,3,4-tetrahydro-naphthalene-1-carboxylic acid, chembl81606, 1-tetralincarboxylic acid, 1,2,3,4-tetrahydro-1-naphthalenecarboxylic acid, 1,2,3,4-tetrahydronaphthalenecarboxylic acid, tetrahydronaphthoic acid
PubChem CID 12598146
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 82°C to 85°C
Quantity 1 g
Solubility Information Soluble in methanol,and chloroform.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.