missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-3-iodobenzene, 98+%, Thermo Scientific™

Product Code. 11414763
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11414763

Brand: Thermo Scientific Alfa Aesar A11647.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2401-21-0
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
MDL Number MFCD00001034
InChI Key VGJKBWPZBVBXGI-UHFFFAOYSA-N
Synonym 2,3-dichloroiodobenzene, 2,3-dichloro-1-iodobenzene, 2,3-dichloro iodobenzene, dichloroiodobenzene, benzene, dichloroiodo, benzene, 1,2-dichloro-3-iodo, 1,2-dichloro-3-iodo-benzene, pubchem3690, 2,3-dichloro-iodobenzene, acmc-209g7t
PubChem CID 75459
IUPAC Name 1,2-dichloro-3-iodobenzene
SMILES C1=CC(=C(C(=C1)I)Cl)Cl

Specifications

Melting Point 33°C to 37°C
Boiling Point 121°C to 123°C (16 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 2247168
Sensitivity Light sensitive
Formula Weight 272.9
Percent Purity ≥98%
Chemical Name or Material 1,2-Dichloro-3-iodobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.