missing translation for 'onlineSavingsMsg'
Learn More

1,2-O-Cyclohexylidene-alpha-D-glucofuranose, 97%, Thermo Scientific™

Product Code. 15408249
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15408249

Brand: Thermo Scientific Alfa Aesar J66761.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16832-21-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00142481
InChI Key KAUCUKASFLXURN-RCZSTQMZSA-N
Synonym r-1-3a'r,5'r,6's,6a'r-6'-hydroxytetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-a-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-, a-d-glucofuranose, a-d-glucofuranose,1,2-o-cyclohexylidene, 1-o,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexane-1,1-diyl-alpha-d-xylo-hexofuranose, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxole-5'-yl ethane-1,2-diol
PubChem CID 2724778
IUPAC Name (1R)-1-[(3aR,5R,6S,6aR)-6-hydroxyspiro[3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole-2,1'-cyclohexane]-5-yl]ethane-1,2-diol
SMILES C1CCC2(CC1)OC3C(C(OC3O2)C(CO)O)O

Specifications

Melting Point 163°C to 168°C
Color White
Quantity 5 g
Physical Form Powder
Formula Weight 260.28
Percent Purity 97%
Chemical Name or Material 1,2-O-Cyclohexylidene-alpha-D-glucofuranose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.