missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluorobenzene, 98+%

Product Code. 11468566
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11468566

Brand: Thermo Scientific Alfa Aesar A15984.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-38-3 | C6H3F3 | 132.085 g/mol

1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-38-3
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00000333
InChI Key JXUKFFRPLNTYIV-UHFFFAOYSA-N
Synonym benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene
PubChem CID 9745
IUPAC Name 1,3,5-trifluorobenzene
SMILES C1=C(C=C(C=C1F)F)F

Specifications

Melting Point -7°C to -4°C
Density 1.279
Boiling Point 75°C to 76°C
Flash Point −7°C (19°F)
Refractive Index 1.415
Quantity 25 g
UN Number UN2387
Beilstein 1906457
Solubility Information Insoluble in water.
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Trifluorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3,5-Trifluorobenzene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,5-Trifluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.