Learn More
1,3,5-Trimethyl-1H-pyrazole, 98%, Thermo Scientific™
75.10 EUR - 248.00 EUR
Chemical Identifiers
CAS | 1072-91-9 |
---|---|
Molecular Formula | C6H10N2 |
Molecular Weight (g/mol) | 110.16 |
MDL Number | MFCD00015536 |
InChI Key | HNOQAFMOBRWDKQ-UHFFFAOYSA-N |
Synonym | 1,3,5-trimethyl-1h-pyrazole, 1h-pyrazole, 1,3,5-trimethyl, pyrazole, 1,3,5-trimethyl, akos pao-1133, pubchem20856, vitas-bb tbb000655, buttpark 15357-40, ksc490o8l, 1,3,5-trimethylpyrazole, 1,3,5-trimethyl-1h-pyrazole # |
PubChem CID | 14081 |
IUPAC Name | 1,3,5-trimethylpyrazole |
SMILES | CC1=CC(=NN1C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1072-91-9 | |
110.16 | |
HNOQAFMOBRWDKQ-UHFFFAOYSA-N | |
14081 | |
CC1=CC(=NN1C)C |
C6H10N2 | |
MFCD00015536 | |
1,3,5-trimethyl-1h-pyrazole, 1h-pyrazole, 1,3,5-trimethyl, pyrazole, 1,3,5-trimethyl, akos pao-1133, pubchem20856, vitas-bb tbb000655, buttpark 15357-40, ksc490o8l, 1,3,5-trimethylpyrazole, 1,3,5-trimethyl-1h-pyrazole # | |
1,3,5-trimethylpyrazole |
Specifications
1072-91-9 | |
168°C to 170°C | |
C6H10N2 | |
UN1325 | |
1,3,5-trimethyl-1h-pyrazole, 1h-pyrazole, 1,3,5-trimethyl, pyrazole, 1,3,5-trimethyl, akos pao-1133, pubchem20856, vitas-bb tbb000655, buttpark 15357-40, ksc490o8l, 1,3,5-trimethylpyrazole, 1,3,5-trimethyl-1h-pyrazole # | |
CC1=CC(=NN1C)C | |
110.16 | |
110.16 | |
1,3,5-Trimethyl-1H-pyrazole |
33°C to 38°C | |
33°C (91°F) | |
MFCD00015536 | |
110515 | |
HNOQAFMOBRWDKQ-UHFFFAOYSA-N | |
1,3,5-trimethylpyrazole | |
14081 | |
98% |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.