missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane, 94%, Thermo Scientific™

Product Code. 11347615
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2943-70-6
Molecular Formula C6H14Cl4OSi2
Molecular Weight (g/mol) 300.147
MDL Number MFCD00040856
InChI Key YMDCHYWOAHMASL-UHFFFAOYSA-N
PubChem CID 76260
IUPAC Name dichloromethyl-[dichloromethyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(C(Cl)Cl)O[Si](C)(C)C(Cl)Cl

Specifications

Boiling Point 149°C to 150°C (40mmHg)
Quantity 25 g
Beilstein 1762720
Flash Point 65°C (149°F)
Synonym 1,3-bis dichloromethyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis dichloromethyl tetramethyldisiloxane, bis dichloromethyl tetramethyldisiloxane, disiloxane, 1,3-bis dichloromethyl-1,1,3,3-tetramethyl, dichloromethyl dichloromethyl dimethylsilyl oxy dimethylsilane, acmc-20ap5a, disiloxane,3-bis dichloromethyl-1,1,3,3-tetramethyl, disiloxane,1,3-bis dichloromethyl-1,1,3,3-tetramethyl, 1,3-bis dichloromethyl-1,1,3,3-tetramethylpropanedisiloxane
Formula Weight 300.16
Percent Purity 94%
Refractive Index 1.466
Density 1.221
Chemical Name or Material 1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'