missing translation for 'onlineSavingsMsg'
Learn More

1,3-diolein, ∼99%, MP Biomedicals™

Product Code. 11491032 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 11491032

Brand: MP Biomedicals 0219515210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2465-32-9
Molecular Formula C39H72O5
Molecular Weight (g/mol) 621
MDL Number MFCD00042913
InChI Key DRAWQKGUORNASA-CLFAGFIQSA-N
Synonym 1,3-diolein, 1,3-dioleoylglycerol, glycerol 1,3-dioleate, unii-erf7s0xx7k, 2-hydroxy-1,3-propanediyl dioleate, erf7s0xx7k, diacylglycerol 18:1/0:0/18:1, 9-octadecenoic acid z-, 2-hydroxy-1,3-propanediyl ester, capmul mpgo, caprol mpgo
PubChem CID 5497165
ChEBI CHEBI:75735
IUPAC Name [2-hydroxy-3-[(Z)-octadec-9-enoyl]oxypropyl] (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)O

Specifications

Color Colorless
Density 0.934g/cm3 (L)
Assay Percent Range ∽ 99%
Quantity 10 mg
Formula Weight 621
Percent Purity 0.994
Physical Form Frozen Liquid
Chemical Name or Material 1, 3-diolein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.