missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals

28.04 EUR - 70.85 EUR

Chemical Identifiers

CAS 106-46-7
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000604
InChI Key OCJBOOLMMGQPQU-UHFFFAOYSA-N
Synonym p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
PubChem CID 4685
ChEBI CHEBI:28618
IUPAC Name 1,4-dichlorobenzene
SMILES C1=CC(=CC=C1Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11414220
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16290.22
100 g
28.04 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
11457747
View Documents
Thermo Scientific Alfa Aesar
A16290.36
500 g
29.04 EUR
500g
Please to purchase this item. Need a web account? Register with us today!
11467747
View Documents
Thermo Scientific Alfa Aesar
A16290.0E
2500 g
70.85 EUR
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
Specifications

Chemical Identifiers

106-46-7
146.998
OCJBOOLMMGQPQU-UHFFFAOYSA-N
4685
1,4-dichlorobenzene
C6H4Cl2
MFCD00000604
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
CHEBI:28618
C1=CC(=CC=C1Cl)Cl

Specifications

106-46-7
1.248
65°C (149°F)
C6H4Cl2
UN3077
14,3057
It is insoluble in water.
C1=CC(=CC=C1Cl)Cl
146.998
CHEBI:28618
≥99%
52°C to 56°C
173°C to 174°C
Strong
MFCD00000604
1680023
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
OCJBOOLMMGQPQU-UHFFFAOYSA-N
1,4-dichlorobenzene
4685
147
1,4-Dichlorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302-H332-H319
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H350

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 203-400-5

missing translation for 'rtecsNumber' : CZ4550000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only