missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine, 97%, Thermo Scientific Chemicals

Product Code. 15429337
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15429337

Brand: Thermo Scientific Alfa Aesar H52577.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 224779-27-5
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD04972246
InChI Key NICJOYHYEDFTIX-UHFFFAOYSA-N
Synonym n-boc-3-hydroxy-1,2,3,6-tetrahydropyridine, tert-butyl 5-hydroxy-5,6-dihydropyridine-1 2h-carboxylate, 1-boc-3-hydroxy-1,2,3,6-tetrahydropyridine, 3-hydroxy-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester, 3-hydroxy-1,2,3,6-tetrahydropyridine, n-boc protected, tert-butyl 5,6-dihydro-5-hydroxypyridine-1 2h-carboxylate, tert-butyl 3-hydroxy-3,6-dihydropyridine-1 2h-carboxylate, tert-butyl 3-hydroxy-1,2,3,6-tetrahydropyridine-1-carboxylate, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-3-hydroxy-, 1,1-dimethylethyl ester, 1-boc-3-hydroxy-1,2,3,6-tetrahydro-pyridine
PubChem CID 17750131
IUPAC Name tert-butyl 3-hydroxy-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC=CC(C1)O

Specifications

Quantity 1 g
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material 1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.