missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-azetidine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.
Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.
Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.
Specifications
Boiling Point | 77°C to 82°C (2mmHg) |
Formula Weight | 185.22 |
Percent Purity | 97% |
Quantity | 250 mg |
Sensitivity | Air sensitive |
Chemical Name or Material | 1-Boc-azetidine-3-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'