Learn More
1-Boc-pyrrole-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
28.04 EUR - 302.51 EUR
Chemical Identifiers
CAS | 1072944-98-9 |
---|---|
Molecular Formula | C15H24BNO4 |
Molecular Weight (g/mol) | 293.17 |
MDL Number | MFCD11504960 |
InChI Key | DZQURIUJAGTAFW-UHFFFAOYSA-N |
Synonym | tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate |
PubChem CID | 46739108 |
IUPAC Name | tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15421259
|
Thermo Scientific Alfa Aesar
H64171.MD |
250 mg | N/A | ||||||
15401259
|
Thermo Scientific Alfa Aesar
H64171.03 |
1 g |
79.09 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15411259
|
Thermo Scientific Alfa Aesar
H64171.06 |
5 g |
302.51 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1072944-98-9 | |
293.17 | |
DZQURIUJAGTAFW-UHFFFAOYSA-N | |
46739108 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C |
C15H24BNO4 | |
MFCD11504960 | |
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate | |
tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate |
Specifications
1072944-98-9 | |
MFCD11504960 | |
Slightly soluble in water. | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=CN2C(=O)OC(C)(C)C | |
293.17 | |
293.17 | |
Solid |
C15H24BNO4 | |
tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, 1-boc-pyrrole-2-boronic acid, pinacol ester, 1-boc-pyrrole-2-boronic acid pinacol ester, t-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate, tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrole-1-carboxylate, pubchem23523, 1-boc-pyrrole-2-boronic acid,pinacol ester, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid pinacol ester, 1-boc-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole, tert-butyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrole-1-carboxylate | |
DZQURIUJAGTAFW-UHFFFAOYSA-N | |
tert-butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrole-1-carboxylate | |
46739108 | |
98% | |
1-Boc-pyrrole-2-boronic acid pinacol ester |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.