missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloroethane, 98%, Thermo Scientific Chemicals

48.54 EUR - 177.89 EUR

Chemical Identifiers

CAS 107-04-0
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.41
MDL Number MFCD00000962
InChI Key IBYHHJPAARCAIE-UHFFFAOYSA-N
Synonym ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
PubChem CID 7849
ChEBI CHEBI:19032
IUPAC Name 1-bromo-2-chloroethane
SMILES C(CBr)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10472211
View Documents Promotion Details
Thermo Scientific Acros
214181000
100 mL Glass bottle
48.54 EUR
100mL
Please to purchase this item. Need a web account? Register with us today!
10616164
View Documents
Thermo Scientific Acros
214185000
500 mL Glass bottle
177.89 EUR
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-04-0
143.41
IBYHHJPAARCAIE-UHFFFAOYSA-N
7849
1-bromo-2-chloroethane
C2H4BrCl
MFCD00000962
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
CHEBI:19032
C(CBr)Cl

Specifications

107-04-0
100.0
Colorless to Yellow
106.0°C to 107.0°C
97.5% min. (GC)
ClCH2CH2Br
MFCD00000962
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
IBYHHJPAARCAIE-UHFFFAOYSA-N
1-bromo-2-chloroethane
7849
143.41
Liquid
97.5
-17.0°C
1.7300g/mL
Authentic
C2H4BrCl
1.4900 to 1.4920
1.73
Solubility in water: 7g/L (20°C). Other solubilities: soluble in alcohol and ether
C(CBr)Cl
143.41
CHEBI:19032
98%
1-Bromo-2-chloroethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-456-

Product Identifier
  • 1-Bromo-2-chloroethane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : 1-Bromo-2-chloroethane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only