Learn More
Integra Biosciences™ 1-Channel Adapter
For use with VACUBOY Hand Operator.
15.75 EUR - 40.60 EUR
Specifications
For Use With (Equipment) VACUBOY Hand Operator
View More Specs
Products 4
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11777312
View Documents
Integra Biosciences™
159027
Adapter with Ejector for GripTips
40.60 EUR
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
10090272
View Documents
Integra Biosciences™
155522
Adapter, 150 mm
24.65 EUR
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
10319341
View Documents
Integra Biosciences™
155502
Adapter, 40 mm
15.75 EUR
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
12964417
View Documents
Integra Biosciences™
159023
Adapter for Disposable Tips
15.75 EUR
Pack of 5
Estimated Shipment: 30-01-2025
Description
One-channel adaptor for use with VACUBOY hand operator. 
The VACUBOY hand operator can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
Specifications
VACUBOY Hand Operator