missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecanol, Reagent grade, 98%, Honeywell™
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 22°C to 26°C |
Color | Colorless |
Density | 0.833g/mL at 25°C |
Boiling Point | 260°C to 262°C |
Flash Point | 121°C (249.8°F) |
Packaging | Poly Bottle |
Linear Formula | CH3(CH2)11OH |
Refractive Index | n20/D 1.442(lit.) |
Quantity | 2.5 L |
UN Number | UN3077 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction