missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)indole, 95%, Thermo Scientific Chemicals

Product Code. 11446016
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31271-90-6
Molecular Formula C15H13NO2S
Molecular Weight (g/mol) 271.33
MDL Number MFCD01165032
InChI Key JNRRPYFLDADLJW-UHFFFAOYSA-N
Synonym 1-tosyl-1h-indole, n-tosylindole, 1-p-toluenesulfonyl indole, 1-4-methylbenzenesulfonyl indole, n-p-toluenesulfonyl indole, 1-4-methylphenyl sulfonyl-1h-indole, 1-tosylindole, 1-p-tolylsulfonyl indole, 1-p-toluenesulphonyl indole, 4-methylphenyl sulfonyl indole
PubChem CID 10956616
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12

Specifications

Melting Point 84°C to 86°C
Quantity 1 g
Solubility Information Insoluble in water.
IUPAC Name 1-(4-methylphenyl)sulfonylindole
Formula Weight 271.33
Percent Purity 95%
Chemical Name or Material 1-(p-Toluenesulfonyl)indole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'