missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15641370
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

Melting Point -78°C
Color Colorless
Density 0.811g/mL at 25°C
Boiling Point 136°C to 138°C
Flash Point 49°C (120.2°F)
Packaging Glass Bottle
Linear Formula CH3(CH2)4OH
Refractive Index n20/D 1.409(lit.)
Quantity 1 L
UN Number UN1105
Beilstein 1730975
Formula Weight 88.15g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material 1-Pentanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.