missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylimidazole, 99%, Thermo Scientific™

Product Code. 11302876
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11302876

Brand: Thermo Scientific Alfa Aesar L10579.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Questo articolo non è restituibile. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-phenylimidazole is a ligand for cytochrome P-450 and used as an inhibitor of microsomal oxidation. 1-Phenylimidazole is a suitable reagent used to investigate its effect on the citrulline formation by bovine brain nitric-oxide synthase. 1-Phenylimidazole is reported to be inhibitor of calmodulin-dependent nitric-oxide synthase from bovine brain and GHs pituitary cells.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7164-98-9
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00041204
InChI Key SEULWJSKCVACTH-UHFFFAOYSA-N
Synonym 1-phenyl-1h-imidazole, phenylimidazole, n-phenylimidazole, 1h-imidazole, 1-phenyl, 1h-imidazole, phenyl, n-phenyl imidazole, pubchem12611, acmc-20aos8, 1-phenylimidazole, 1-phenyl-1h-imidazole #
PubChem CID 81595
SMILES C1=CN(C=N1)C1=CC=CC=C1

Specifica

Melting Point 13°C
Density 1.14
Boiling Point 142°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6
Quantity 250 mg
Beilstein 112356
Solubility Information Slightly soluble in water.
IUPAC Name 1-phenylimidazole
Formula Weight 144.18
Percent Purity 99%
Chemical Name or Material 1-Phenylimidazole
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato