missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecyne, 97%, Thermo Scientific Chemicals

Product Code. 11327725
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26186-02-7
Molecular Formula C13H24
Molecular Weight (g/mol) 180.34
MDL Number MFCD00015074
InChI Key GZEDKDBFUBPZNG-UHFFFAOYSA-N
Synonym 1-tridecyne, tridecyne, acmc-1cinm
PubChem CID 117754
IUPAC Name tridec-1-yne
SMILES CCCCCCCCCCCC#C

Specifications

Density 0.78
Boiling Point 116°C to 118°C (10 mmHg)
Flash Point 75°C (167°F)
Linear Formula CH3(CH2)10C≡CH
Refractive Index 1.438
Quantity 1 g
Beilstein 1744658
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 180.34
Percent Purity 97%
Chemical Name or Material 1-Tridecyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'