missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Calcitonin R Protein
Shop All Bio Techne Products
Click to view available options
:
10 μg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Immunoaffinity Purification |
| Formulation | 25 mM Tris-HCl pH8.0 in 2% glycerol. |
| Gene ID (Entrez) | 799 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 55.3 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | calcitonin receptor, CRT, CTR, CT-R, CTR1 |
| Gene Symbol | CALCR |
| Research Category | Cancer, GPCR, Signal Transduction |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction