Learn More
Helios Italquartz 1234 DIN Standard Crucibles, Transparent, Ridge Points
1234 DIN standard crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 63P00105
This item is not returnable.
View return policy
Description
- Tubular shape with supporting ridge points
- Transparent
- Conform to 1234 DIN Standard
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Transparent | |
10 cc | |
38 mm |
Quartz | |
25 mm | |
1234 DIN standard crucible |