Learn More
Helios Italquartz 1234 DIN Standard Crucibles, Transparent, Ridge Points
1234 DIN standard crucibles constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 63P00105
Product Code. 15888044
  • 91.85 EUR / Each
Estimated Shipment: 19-02-2025
Description
  • Tubular shape with supporting ridge points
  • Transparent
  • Conform to 1234 DIN Standard
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
10 cc
38 mm
Quartz
25 mm
1234 DIN standard crucible