Learn More
Axygen™ 1250μL Universal Pipette Tips
GREENER_CHOICE
Product Code. 12531308 Shop All Axygen Scientific Products
Click to view available options
Packaging:
Bulk
Rack
Unit Size:
Pack of 1152
Pack of 3456
Pack of 500
Pack of 5000
Product Code. 12531308
Brand: Axygen™ MTX1250C
Only null left
Add to basket
Add to basket
Matrix compatible tips in clear color
  • Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 15 to 1250μL
  • Available in bulk or racked formats
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1250 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Certifications/Compliance RNase, DNase-free
Packaging Bulk
For Use With (Equipment) Thermo Scientific Matrix Impact