Learn More
14-Heptacosanone, 97%
Product Code. 11449636
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11449636
Brand: Thermo Scientific Alfa Aesar B20191.06
Request Bulk or Custom Format
CAS: 542-50-7 | C27H54O | 394.728 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 542-50-7
Molecular Formula C27H54O
Molecular Weight (g/mol) 394.728
MDL Number MFCD00026598
InChI Key VCZMOZVQLARCOE-UHFFFAOYSA-N
Synonym 14-heptacosanone, ditridecyl ketone, myristone, unii-fq5vc928nn, fq5vc928nn, acmc-209leu, 14-heptacosanon, di-n-tridecyl ketone
PubChem CID 10955
IUPAC Name heptacosan-14-one
SMILES CCCCCCCCCCCCCC(=O)CCCCCCCCCCCCC
Specifications
Melting Point 76°C to 79°C
Quantity 5 g
Beilstein 1796794
Formula Weight 394.73
Percent Purity 97%
Chemical Name or Material 14-Heptacosanone