missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1422A Tyvek™ Autoclave Bags
BIOPHARMAGREENER_CHOICE

1422A Tyvek™ Autoclave Bags with drawstring closure create a superior microbial barrier

1845.00 EUR - 5220.00 EUR

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type String Closure
Color White
For Use With (Application) Steam Sterilization
View More Specs

Products 15
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
17737887
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A0314D
3 x 14 in.
1960.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17747877
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A0212
2 x 12 in.
2610.00 EUR
Pack of 500
Add to basket
 
17748017
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A1526D
15 x 26 in.
3110.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17737907
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A0512D
5 x 12 in.
2005.00 EUR
Pack of 250
Add to basket
 
17747907
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A0515D
5 x 15 in.
2140.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17757937
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A1007D
10 x 7 in.
1845.00 EUR
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
17767947
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A1026D
10 x 26 in.
2725.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17737957
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A1062
10 x 62 in.
3260.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17707997
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A1246
12 x 46 in.
3040.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17748027
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A1622D
16 x 22 in.
3220.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17708097
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A2432D
24 x 32 in.
3940.00 EUR
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
17718107
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A2462
24 x 62 in.
5220.00 EUR
Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17718127
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A3036
30 x 36 in.
3735.00 EUR
Pack of 250
Add to basket
 
17718167
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A4242
42 x 42 in.
2365.00 EUR
Pack of 125
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17758177
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
BGT2A5054
50 x 54 in.
2395.00 EUR
Pack of 100
Add to basket
 
Description

Description

  • Manufactured and packaged in a controlled cleanroom environment
  • Designed for steam sterilization of large, or oddly shaped pharmaceutical equipment
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Closed via the adjustable Tyvek™ drawstring closure system
  • Eliminates the need to cut material or removal of particulate generating autoclave tape
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
Specifications

Specifications

Autoclavable
String Closure
Steam Sterilization
250/Pk.
Certificate of Conformance (COC)
White
HDPE
Videos
SDS
Documents

Documents

Product Certifications