Learn More
Applied Biosystems™ 3130xl/3100 Genetic Analyzer 16-Capillary Array, 50cm
Use for fragment analysis and rapid sequencing applications.
Brand:  Applied Biosystems™ 4315930
Product Code. 15771816
Description
Description
  • For the 3130xl and 3100 Genetic Analyzer
  • Usage per array is 100 runs (with use of Applied Biosystems reagents).
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 cm
16-Capillary Array, 50cm Capillary Length, 61cm x 50μm I.D. Capillary, 165 min. Run Time, Use for Short Fragment Analysis, Room Temperature, For Use With 3130xl/3100 Genetic Analyzer
Sequencing, Fragment Analysis
Genetic Analyzer
Room Temperature
Store the array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to your genetic analyser's maintenance, troubleshooting and reference guide.
We guarantee the performance of the 3130 Capillary arrays up to 100 runs and within 1 year of purchase.
3100 Genetic Analyzer, 3130xl Genetic Analyzer
16 Capillaries
1 Array
165 min