Learn More
Bochem™ 18/10 Stainless Steel Chemical Spoon
Product Code. 10300681
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
32 mm
40 mm
45 mm
48 mm
Diameter (Metric) Outer:
1 mm
2 mm
5 mm
Unit Size:
Each
Pack of 10
Product Code. 10300681
Brand: Bochem™ 3454
Stainless steel spoon for scooping chemicals.
  • Chemical spoon
  • With spatula handle
Specifications
Quantity 1 Ea.
Product Type Chemical Scoop
Length (Metric) Blade 48 mm
Width (Metric) 35 mm
Diameter (Metric) Outer 5 mm
Material Stainless Steel (18/10)
Length (Metric) 250 mm