Learn More
Bochem™ 18/10 Stainless Steel Crucible Tongs
Click to view available options
Length (Metric):
200 mm
250 mm
300 mm
Unit Size:
Each
Specifications
Specifications
| Length (Metric) | 200 mm |
| Material | Stainless Steel (18/10) |
| Type | Crucible Tongs |