missing translation for 'onlineSavingsMsg'
Learn More

(1R,4S)-(+)-4-Aminocyclopent-2-enecarboxylic acid, 95%, Thermo Scientific™

Product Code. 15429505
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(1R,4S)-(+)-4-Aminocyclopent-2-enecarboxylic acid is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134003-04-6
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00211285
InChI Key VTCHZFWYUPZZKL-CRCLSJGQSA-N
Synonym 1r,4s-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-aminocyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-aminocyclopent-2-enecarboxylic acid, +-1r,4s-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-amino-2-cyclopentene-1-carboxylic acid, 1r,4s-4-amino-cyclopent-2-ene carboxylic acid, 1r,4s-+-4-aminocyclopent-2-enecarboxylic acid, 2-cyclopentene-1-carboxylic acid, 4-amino-, 1r,4s, pubchem18728, +-1r,4s-g-homocycloleu-2-ene
PubChem CID 2734525
IUPAC Name (1R,4S)-4-aminocyclopent-2-ene-1-carboxylic acid
SMILES C1C(C=CC1N)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 127.14
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (1R,4S)-(+)-4-Aminocyclopent-2-enecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'