missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, ≥99.5%, Honeywell™
181.28 EUR - 1649.07 EUR
Chemical Identifiers
CAS | 540-84-1 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
MDL Number | MFCD00008943 |
InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
PubChem CID | 10907 |
ChEBI | CHEBI:62805 |
IUPAC Name | 2,2,4-trimethylpentane |
SMILES | CC(C)CC(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15616110
|
Honeywell-Chromasolv
650439-1L |
1 L | Glass Bottle |
181.28 EUR
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15626110
|
Honeywell Riedel-de Haën
650439-20L |
20 L | Glass Bottle | N/A | |||||
Description
Chemical Identifiers
540-84-1 | |
114.232 | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
10907 | |
2,2,4-trimethylpentane |
C8H18 | |
MFCD00008943 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CHEBI:62805 | |
CC(C)CC(C)(C)C |
Specifications
540-84-1 | |
0.06 | |
C8H18 | |
MFCD00008943 | |
UN1262 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CC(C)CC(C)(C)C | |
114.232 | |
CHEBI:62805 | |
≥99.5% |
0.692g/mL at 25°C | |
745°C | |
(CH3)2CHCH2C(CH3)3 | |
41mmHg (21°C) | |
1696876 | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
2,2,4-trimethylpentane | |
10907 | |
114.23g/mol | |
2,2,4-Trimethylpentane |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
missing translation for 'dotInformation' : class 3 - PG 2 - Octanes
missing translation for 'rtecsNumber' : 208-759-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 2,2,4-Trimethylpentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2,2,4-trimethylpentane
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title