missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, For pesticide residue analysis, Honeywell Riedel-de Haën™

CHROMASOLV™, for pesticide residue analysis

107.68 EUR - 560.99 EUR

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15684230
View Documents
Honeywell-Chromasolv
34499-1L
1 L
107.68 EUR
1 L
Please to purchase this item. Need a web account? Register with us today!
15694230
View Documents
Honeywell-Chromasolv
34499-2.5L
2.5 L
217.65 EUR
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15604240
View Documents
Honeywell-Chromasolv
34499-6X1L
6 x 1 L
560.99 EUR
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Chemical Identifiers

540-84-1
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
2,2,4-trimethylpentane
C8H18
MFCD00008943
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CHEBI:62805
CC(C)CC(C)(C)C

Specifications

540-84-1
0.06
745°F
(CH3)2CHCH2C(CH3)3
41 mmHg (21°C)
1696876
NHTMVDHEPJAVLT-UHFFFAOYSA-N
2,2,4-trimethylpentane
10907
114.23g/mol
CHROMASOLV™
0.692g/mL at 25°C
−12°C (10.4°F)
C8H18
MFCD00008943
UN1262
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CC(C)CC(C)(C)C
114.232
CHEBI:62805
≥99% (GC)
2,2,4-Trimethylpentane
Videos
Safety and Handling

Safety and Handling

P210-P261-P273-P301 + P310-P331-P501

missing translation for 'rtecsNumber' : SA3320000

Product Identifier
  • 2,2,4-Trimethylpentane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.